Assay Details
Gene Symbol: Zasp66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169911.1 | NP_001163382.1 | 7 - 8 | 1242 | 71 | |
NM_001169913.1 | NP_001163384.1 | 7 - 8 | 1154 | 71 | ||
NM_001169914.3 | NP_001163385.1 | 7 - 8 | 1253 | 71 | ||
NM_001169915.2 | NP_001163386.1 | 6 - 7 | 1093 | 71 | ||
NM_139989.4 | NP_648246.3 | 6 - 7 | 1082 | 71 | ||
NM_168283.4 | NP_729395.2 | 8 - 9 | 1325 | 71 | ||
NM_168284.3 | NP_729396.2 | 8 - 9 | 1314 | 71 | ||
NM_168286.4 | NP_729398.1 | 7 - 8 | 812 | 71 | ||
NM_176303.5 | NP_788481.1 | 6 - 7 | 740 | 71 | ||
NM_176304.5 | NP_788482.1 | 5 - 6 | 580 | 71 | ||
NM_176306.3 | NP_788484.2 | 6 - 7 | 952 | 71 | ||
GenBank mRNA | AY058415.1 | - | 8 - 9 | 1292 | 71 | |
BT029127.1 | - | 7 - 8 | 1222 | 71 | ||
BT088849.1 | - | 7 - 8 | 788 | 71 | ||
BT089012.1 | - | 5 - 6 | 575 | 71 | ||
BT133470.1 | - | 7 - 8 | 1251 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
