Assay Details
Gene Symbol: eIF4E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274697.1 | NP_001261626.1 | 4 - 5 | 884 | 79 | |
NM_001274698.1 | NP_001261627.1 | 3 - 4 | 1185 | 79 | ||
NM_080090.3 | NP_524829.1 | 3 - 4 | 876 | 79 | ||
NM_168333.2 | NP_729480.1 | 2 - 3 | 548 | 79 | ||
NM_168334.1 | NP_729481.1 | 3 - 4 | 822 | 79 | ||
NM_168335.2 | NP_729482.1 | 3 - 4 | 1115 | 79 | ||
NM_168336.2 | NP_729483.1 | 3 - 4 | 916 | 79 | ||
NM_168337.2 | NP_729484.1 | 3 - 4 | 844 | 79 | ||
NM_168338.2 | NP_729485.1 | 3 - 4 | 750 | 79 | ||
GenBank mRNA | BT012467.1 | - | 2 - 3 | 546 | 79 | |
BT044219.1 | - | 3 - 4 | 1090 | 79 | ||
U16139.1 | - | 2 - 3 | 595 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
