Assay Details
| Gene Symbol: eIF4E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274697.1 | NP_001261626.1 | 6 - 7 | 1081 | 60 | |
| NM_001274698.1 | NP_001261627.1 | 5 - 6 | 1382 | 60 | ||
| NM_080090.3 | NP_524829.1 | 5 - 6 | 1073 | 60 | ||
| NM_168333.2 | NP_729480.1 | 4 - 5 | 745 | 60 | ||
| NM_168334.1 | NP_729481.1 | 5 - 6 | 1019 | 60 | ||
| NM_168335.2 | NP_729482.1 | 5 - 6 | 1312 | 60 | ||
| NM_168336.2 | NP_729483.1 | 5 - 6 | 1113 | 60 | ||
| NM_168337.2 | NP_729484.1 | 5 - 6 | 1041 | 60 | ||
| NM_168338.2 | NP_729485.1 | 5 - 6 | 947 | 60 | ||
| GenBank mRNA | BT012467.1 | - | 4 - 5 | 743 | 60 | |
| BT044219.1 | - | 5 - 6 | 1287 | 60 | ||
| U16139.1 | - | 4 - 5 | 792 | 60 | ||
More Information
Set Membership: |
|

eIF4E1
Genomic Map