Assay Details
| Gene Symbol: vnc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140121.2 | NP_648378.1 | 3 | 959 | 83 | |
| NM_168390.2 | NP_729581.1 | 2 | 879 | 83 | ||
| NM_168391.2 | NP_729582.1 | 3 | 1000 | 83 | ||
| NM_168392.2 | NP_729583.1 | 2 | 1039 | 83 | ||
| NM_168393.2 | NP_729584.1 | 1 | 1131 | 83 | ||
| GenBank mRNA | AY061279.1 | - | 2 | 864 | 83 | |
| Gene Symbol: CG42455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169933.2 | NP_001163404.1 | 2 | 1039 | 83 | |
| NM_001169934.2 | NP_001163405.1 | 3 | 959 | 83 | ||
| NM_001169935.2 | NP_001163406.1 | 3 | 1000 | 83 | ||
| GenBank mRNA | BT100174.1 | - | 3 | 1031 | 83 | |
More Information
Set Membership: |
|

vnc
Genomic Map