Assay Details
Gene Symbol: Alg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_168449.2 | NP_729680.1 | 2 - 3 | 727 | 94 | |
GenBank mRNA | AJ431376.1 | - | 2 - 3 | 722 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
