Assay Details
Gene Symbol: pie | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273419.1 | NP_001260348.1 | 1 - 2 | 253 | 58 | |
NM_080001.4 | NP_524740.2 | 1 - 2 | 253 | 58 | ||
GenBank mRNA | AY061141.1 | - | 1 - 2 | 196 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
