Assay Details
Gene Symbol: Rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272879.1 | NP_001259808.1 | 1 - 2 | 147 | 57 | |
NM_080010.4 | NP_524749.1 | 1 - 2 | 147 | 57 | ||
GenBank mRNA | AY075315.1 | - | 1 - 2 | 93 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
