Assay Details
Gene Symbol: GluRIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274692.1 | NP_001261621.2 | 8 - 9 | 2058 | 76 | |
NM_001274693.1 | NP_001261622.1 | 8 - 9 | 2058 | 76 | ||
NM_001316480.1 | NP_001303409.1 | 8 - 9 | 2058 | 76 | ||
NM_080016.4 | NP_524755.2 | 8 - 9 | 2058 | 76 | ||
GenBank mRNA | BT150463.1 | - | 8 - 9 | 2057 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
