Assay Details
Gene Symbol: san | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080040.5 | NP_524779.1 | 2 - 3 | 457 | 62 | |
GenBank mRNA | AF225902.1 | - | 2 - 3 | 458 | 62 | |
AY070826.1 | - | 2 - 3 | 465 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
