Assay Details
| Gene Symbol: LPCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042800.3 | NP_001036265.1 | 7 - 8 | 1796 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPCAT| Gene Symbol: LPCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042800.3 | NP_001036265.1 | 7 - 8 | 1796 | 76 | |
Set Membership: |
|