Assay Details
Gene Symbol: CG2186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272504.1 | NP_001259433.1 | 6 - 7 | 2686 | 89 | |
NM_001382048.1 | NP_001368978.1 | 5 - 6 | 2881 | 89 | ||
NM_132432.2 | NP_572660.1 | 6 - 7 | 2686 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
