Assay Details
Gene Symbol: CDK2AP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272510.1 | NP_001259439.1 | 1 | 1785 | 87 | |
NM_132460.3 | NP_572688.2 | 1 | 1785 | 87 | ||
GenBank mRNA | BT044505.1 | - | 1 | 1762 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
