Assay Details
Gene Symbol: Lint-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272511.1 | NP_001259440.1 | 3 - 4 | 2085 | 72 | |
NM_001272512.1 | NP_001259441.1 | 3 - 4 | 2175 | 72 | ||
NM_001298184.1 | NP_001285113.1 | 3 - 4 | 2052 | 72 | ||
NM_132464.3 | NP_572692.1 | 3 - 4 | 1931 | 72 | ||
GenBank mRNA | AY071333.1 | - | 3 - 4 | 1926 | 72 | |
BT089001.1 | - | 3 - 4 | 1786 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
