Assay Details
| Gene Symbol: ATP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272537.2 | NP_001259466.1 | 1 - 2 | 1300 | 59 | |
| NM_132528.3 | NP_572756.3 | 1 - 2 | 1300 | 59 | ||
More Information
Set Membership: |
|

ATP7
Genomic Map