Assay Details
| Gene Symbol: Aven | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298233.1 | NP_001285162.1 | 1 - 2 | 167 | 83 | |
| NM_132589.2 | NP_572817.1 | 1 - 2 | 125 | 83 | ||
| GenBank mRNA | AY071046.1 | - | 1 - 2 | 125 | 83 | |
More Information
Set Membership: |
|

Aven
Genomic Map