Assay Details
Gene Symbol: Syt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272584.1 | NP_001259513.1 | 5 - 6 | 2216 | 58 | |
NM_132650.2 | NP_572878.1 | 5 - 6 | 2219 | 58 | ||
GenBank mRNA | BT133177.1 | - | 5 - 6 | 2217 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
