Assay Details
| Gene Symbol: AMPdeam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132703.3 | NP_572931.1 | 1 - 2 | 992 | 69 | |
| GenBank mRNA | AY051429.1 | - | 1 - 2 | 992 | 69 | |
More Information
Set Membership: |
|

AMPdeam
Genomic Map