Assay Details
| Gene Symbol: Scamp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272643.1 | NP_001259572.1 | 3 - 4 | 612 | 72 | |
| NM_132795.4 | NP_573023.2 | 3 - 4 | 612 | 72 | ||
| NM_167442.2 | NP_727850.1 | 3 - 4 | 1034 | 72 | ||
| GenBank mRNA | AF241834.1 | - | 3 - 4 | 505 | 72 | |
| AY075562.1 | - | 3 - 4 | 619 | 72 | ||
More Information
Set Membership: |
|

Scamp
Genomic Map