Assay Details
| Gene Symbol: Nprl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132946.3 | NP_573174.1 | 1 - 2 | 298 | 66 | |
| GenBank mRNA | AY051914.1 | - | 1 - 2 | 258 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nprl2| Gene Symbol: Nprl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_132946.3 | NP_573174.1 | 1 - 2 | 298 | 66 | |
| GenBank mRNA | AY051914.1 | - | 1 - 2 | 258 | 66 | |
Set Membership: |
|