Assay Details
| Gene Symbol: Ctl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274482.1 | NP_001261411.1 | 8 - 9 | 2002 | 132 | |
| NM_139627.2 | NP_647884.1 | 8 - 9 | 2017 | 132 | ||
| GenBank mRNA | BT150374.1 | - | 8 - 9 | 2015 | 132 | |
More Information
Set Membership: |
|

Ctl1
Genomic Map