Assay Details
| Gene Symbol: Cip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104031.2 | NP_001097501.1 | 5 - 6 | 640 | 65 | |
| NM_001259687.2 | NP_001246616.1 | 5 - 6 | 528 | 65 | ||
| NM_001259688.2 | NP_001246617.1 | 5 - 6 | 528 | 65 | ||
| NM_001259689.1 | NP_001246618.1 | 4 - 5 | 414 | 65 | ||
| NM_001259690.2 | NP_001246619.1 | 5 - 6 | 528 | 65 | ||
| NM_001259691.2 | NP_001246620.1 | 5 - 6 | 528 | 65 | ||
| NM_001259692.1 | NP_001246621.1 | 5 - 6 | 640 | 65 | ||
| NM_001259693.2 | NP_001246622.1 | 5 - 6 | 528 | 65 | ||
| NM_001274483.1 | NP_001261412.1 | 5 - 6 | 528 | 65 | ||
| NM_001274484.1 | NP_001261413.1 | 5 - 6 | 528 | 65 | ||
| NM_139636.4 | NP_647893.2 | 5 - 6 | 528 | 65 | ||
| GenBank mRNA | BT050431.1 | - | 5 - 6 | 546 | 65 | |
More Information
Set Membership: |
|

Cip4
Genomic Map