Assay Details
Gene Symbol: kri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274522.1 | NP_001261451.1 | 1 - 2 | 346 | 67 | |
NM_001274523.1 | NP_001261452.1 | 1 - 2 | 346 | 67 | ||
NM_139733.2 | NP_647990.1 | 1 - 2 | 346 | 67 | ||
GenBank mRNA | AY069484.1 | - | 1 - 2 | 336 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
