Assay Details
| Gene Symbol: eIF4E4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139795.3 | NP_648052.1 | 3 - 4 | 661 | 60 | |
| GenBank mRNA | BT021432.1 | - | 3 - 4 | 653 | 60 | |
More Information
Set Membership: |
|

eIF4E4
Genomic Map