Assay Details
| Gene Symbol: form3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014571.1 | NP_001014571.1 | 2 - 3 | 981 | 73 | |
| NM_001104057.1 | NP_001097527.1 | 2 - 3 | 981 | 73 | ||
More Information
Set Membership: |
|

form3
Genomic Map