Assay Details
| Gene Symbol: RpL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274572.1 | NP_001261501.1 | 2 - 3 | 178 | 63 | |
| NM_001300037.1 | NP_001286966.1 | 2 - 3 | 116 | 63 | ||
| NM_139834.3 | NP_648091.1 | 3 - 4 | 146 | 63 | ||
| GenBank mRNA | AY113554.1 | - | 3 - 4 | 145 | 63 | |
| BT030448.1 | - | 2401 | 63 | |||
More Information
Set Membership: |
|

RpL18
Genomic Map