Assay Details
| Gene Symbol: unc-13-4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274575.2 | NP_001261504.1 | 6 - 7 | 1687 | 82 | |
| NM_139839.5 | NP_648096.4 | 6 - 7 | 1687 | 82 | ||
More Information
Set Membership: |
|

unc-13-4A
Genomic Map