Assay Details
Gene Symbol: CG43163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259744.1 | NP_001246673.1 | 6 - 7 | 5267 | 79 | |
NM_139971.2 | NP_648228.2 | 5 - 6 | 5175 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
