Assay Details
| Gene Symbol: CG43163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259744.1 | NP_001246673.1 | 7 - 8 | 6034 | 77 | |
| NM_139971.2 | NP_648228.2 | 6 - 7 | 5942 | 77 | ||
More Information
Set Membership: |
|

CG43163
Genomic Map