Assay Details
| Gene Symbol: GstO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139980.3 | NP_648237.1 | 2 - 3 | 668 | 112 | |
| GenBank mRNA | AY071499.1 | - | 2 - 3 | 663 | 112 | |
More Information
Set Membership: |
|

GstO1
Genomic Map