Assay Details
| Gene Symbol: ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169481.2 | NP_001162952.1 | 7 - 8 | 2308 | 64 | |
| NM_001273450.1 | NP_001260379.1 | 5 - 6 | 1958 | 64 | ||
| NM_057214.5 | NP_476562.1 | 7 - 8 | 2360 | 64 | ||
| NM_057215.5 | NP_476563.1 | 7 - 8 | 2356 | 64 | ||
More Information
Set Membership: |
|

ab
Genomic Map