Assay Details
| Gene Symbol: Gpdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273182.1 | NP_001260111.1 | 3 - 4 | 893 | 73 | |
| NM_001273183.2 | NP_001260112.1 | 3 - 4 | 893 | 73 | ||
| NM_001273184.2 | NP_001260113.1 | 3 - 4 | 893 | 73 | ||
| NM_057217.4 | NP_476565.1 | 3 - 4 | 571 | 73 | ||
| NM_057218.4 | NP_476566.1 | 3 - 4 | 571 | 73 | ||
| NM_057219.4 | NP_476567.1 | 3 - 4 | 571 | 73 | ||
| GenBank mRNA | BT044153.1 | - | 3 - 4 | 557 | 73 | |
More Information
Set Membership: |
|

Gpdh1
Genomic Map