Assay Details
| Gene Symbol: Gpdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273183.2 | NP_001260112.1 | 6 - 7 | 1510 | 85 | |
| NM_057217.4 | NP_476565.1 | 6 - 7 | 1188 | 85 | ||
| GenBank mRNA | BT044153.1 | - | 6 - 7 | 1174 | 85 | |
More Information
Set Membership: |
|

Gpdh1
Genomic Map