Assay Details
| Gene Symbol: ImpE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057232.5 | NP_476580.1 | 1 | 1539 | 90 | |
| GenBank mRNA | AY061270.1 | - | 1 | 1503 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ImpE2| Gene Symbol: ImpE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057232.5 | NP_476580.1 | 1 | 1539 | 90 | |
| GenBank mRNA | AY061270.1 | - | 1 | 1503 | 90 | |
Set Membership: |
|