Assay Details
| Gene Symbol: esg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057252.4 | NP_476600.1 | 1 | 1916 | 151 | |
| GenBank mRNA | BT133380.1 | - | 1 | 1913 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

esg| Gene Symbol: esg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057252.4 | NP_476600.1 | 1 | 1916 | 151 | |
| GenBank mRNA | BT133380.1 | - | 1 | 1913 | 151 | |
Set Membership: |
|