Assay Details
| Gene Symbol: crp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299047.1 | NP_001285976.1 | 2 - 3 | 942 | 76 | |
| NM_057257.4 | NP_476605.1 | 2 - 3 | 942 | 76 | ||
| GenBank mRNA | AF158371.1 | - | 2 - 3 | 912 | 76 | |
| BT053689.1 | - | 2 - 3 | 940 | 76 | ||
| BT058007.1 | - | 2 - 3 | 940 | 76 | ||
More Information
Set Membership: |
|

crp
Genomic Map