Assay Details
| Gene Symbol: AspRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274001.1 | NP_001260930.1 | 2 - 3 | 121 | 114 | |
| NM_001274002.2 | NP_001260931.1 | 3 - 4 | 468 | 114 | ||
| NM_057261.4 | NP_476609.1 | 2 - 3 | 198 | 114 | ||
| GenBank mRNA | AY070532.1 | - | 2 - 3 | 163 | 114 | |
More Information
Set Membership: |
|

AspRS
Genomic Map