Assay Details
| Gene Symbol: AspRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274001.1 | NP_001260930.1 | 3 - 4 | 448 | 87 | |
| NM_001274002.2 | NP_001260931.1 | 4 - 5 | 795 | 87 | ||
| NM_057261.4 | NP_476609.1 | 3 - 4 | 525 | 87 | ||
| GenBank mRNA | AY070532.1 | - | 3 - 4 | 490 | 87 | |
More Information
Set Membership: |
|

AspRS
Genomic Map