Assay Details
Gene Symbol: Mal-A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057277.3 | NP_476625.2 | 1 - 2 | 1525 | 73 | |
GenBank mRNA | AY071566.1 | - | 1 - 2 | 1519 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
