Assay Details
Gene Symbol: Appl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258519.1 | NP_001245448.1 | 8 - 9 | 1732 | 64 | |
NM_001258520.2 | NP_001245449.1 | 7 - 8 | 1575 | 64 | ||
NM_001258521.2 | NP_001245450.1 | 8 - 9 | 1749 | 64 | ||
NM_001258522.2 | NP_001245451.1 | 8 - 9 | 1734 | 64 | ||
NM_001258523.2 | NP_001245452.1 | 8 - 9 | 1749 | 64 | ||
NM_057278.5 | NP_476626.2 | 8 - 9 | 1746 | 64 | ||
GenBank mRNA | AF181628.1 | - | 7 - 8 | 1721 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
