Assay Details
| Gene Symbol: Appl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258519.1 | NP_001245448.1 | 10 - 11 | 2561 | 81 | |
| NM_001258520.2 | NP_001245449.1 | 9 - 10 | 2407 | 81 | ||
| NM_001258521.2 | NP_001245450.1 | 10 - 11 | 2581 | 81 | ||
| NM_001258522.2 | NP_001245451.1 | 10 - 11 | 2566 | 81 | ||
| NM_001258523.2 | NP_001245452.1 | 10 - 11 | 2581 | 81 | ||
| NM_057278.5 | NP_476626.2 | 10 - 11 | 2578 | 81 | ||
| GenBank mRNA | AF181628.1 | - | 9 - 10 | 2553 | 81 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				

Appl
Genomic Map