Assay Details
Gene Symbol: Mal-A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057280.4 | NP_476628.2 | 3 - 4 | 1698 | 93 | |
GenBank mRNA | AY119654.1 | - | 3 - 4 | 1691 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
