Assay Details
| Gene Symbol: PpY-55A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299625.1 | NP_001286554.1 | 1 | 987 | 76 | |
| NM_057341.4 | NP_476689.1 | 1 | 987 | 76 | ||
| GenBank mRNA | AY075165.1 | - | 1 | 987 | 76 | |
More Information
Set Membership: |
|

PpY-55A
Genomic Map