Assay Details
| Gene Symbol: sna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057384.4 | NP_476732.1 | 1 | 1531 | 168 | |
| GenBank mRNA | BT010255.1 | - | 1 | 1525 | 168 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sna| Gene Symbol: sna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057384.4 | NP_476732.1 | 1 | 1531 | 168 | |
| GenBank mRNA | BT010255.1 | - | 1 | 1525 | 168 | |
Set Membership: |
|