Assay Details
Gene Symbol: alpha-Spec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274358.1 | NP_001261287.1 | 6 - 7 | 6897 | 77 | |
NM_057391.4 | NP_476739.1 | 5 - 6 | 6801 | 77 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
