Assay Details
| Gene Symbol: Su(dx) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272969.1 | NP_001259898.1 | 3 - 4 | 661 | 62 | |
| NM_001272970.1 | NP_001259899.1 | 3 - 4 | 480 | 62 | ||
| NM_057405.3 | NP_476753.1 | 3 - 4 | 661 | 62 | ||
| NM_164448.2 | NP_722753.1 | 3 - 4 | 877 | 62 | ||
| NM_164449.2 | NP_722754.1 | 3 - 4 | 438 | 62 | ||
| GenBank mRNA | AF152865.1 | - | 3 - 4 | 572 | 62 | |
| BT021390.1 | - | 3 - 4 | 654 | 62 | ||
More Information
Set Membership: |
|

Su(dx)
Genomic Map