Assay Details
Gene Symbol: Top2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273651.1 | NP_001260580.1 | 3 - 4 | 1080 | 102 | |
NM_057412.5 | NP_476760.1 | 2 - 3 | 825 | 102 | ||
GenBank mRNA | BT150455.1 | - | 2 - 3 | 820 | 102 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
