Assay Details
Gene Symbol: Top2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273651.1 | NP_001260580.1 | 5 - 6 | 4540 | 69 | |
NM_057412.5 | NP_476760.1 | 4 - 5 | 4285 | 69 | ||
GenBank mRNA | BT150455.1 | - | 4 - 5 | 4280 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
