Assay Details
| Gene Symbol: tor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103742.2 | NP_001097212.2 | 10 - 11 | 2509 | 68 | |
| NM_057414.4 | NP_476762.1 | 10 - 11 | 2524 | 68 | ||
| GenBank mRNA | AY071403.1 | - | 10 - 11 | 2522 | 68 | |
More Information
Set Membership: |
|

tor
Genomic Map