Assay Details
| Gene Symbol: tor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103742.2 | NP_001097212.2 | 4 - 5 | 940 | 76 | |
| NM_057414.4 | NP_476762.1 | 4 - 5 | 940 | 76 | ||
| GenBank mRNA | AY071403.1 | - | 4 - 5 | 938 | 76 | |
More Information
Set Membership: |
|

tor
Genomic Map