Assay Details
| Gene Symbol: RpS27A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057430.4 | NP_476778.1 | 2 - 3 | 379 | 64 | |
| GenBank mRNA | BT001653.1 | - | 1129 | 64 | ||
| BT099554.1 | - | 1 - 2 | 359 | 64 | ||
| M22536.1 | - | 2 - 3 | 457 | 64 | ||
More Information
Set Membership: |
|

RpS27A
Genomic Map